Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 6622 |
Gene Name: | synuclein alpha |
Gene Aliases: |
NACP, PARK1, PARK4, PD1 |
Location: |
Chr.4:89724099-89838324 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SNCA | NM_000345.3 | 6 | 1442 | NP_000336.1 |
NM_001146054.1 | 6 | 1438 | NP_001139526.1 | |
NM_001146055.1 | 6 | 1249 | NP_001139527.1 | |
NM_007308.2 | 5 | 1354 | NP_009292.1 | |
XM_011532208.2 | 6 | 1319 | XP_011530510.1 | |
XM_017008562.1 | 6 | 1313 | XP_016864051.1 | |
XM_017008563.1 | 6 | 1339 | XP_016864052.1 | |
BC108275.1 | 6 | 1396 | AAI08276.1 | |
D31839.1 | BAA06625.1 | |||
JN709859.1 | 5 | 1396 | ||
JN709860.1 | 4 | 1312 | ||
JN709861.1 | 5 | 1358 | ||
JN709862.1 | 5 | 1400 | ||
JN709863.1 | 4 | 1316 | ||
JN709864.1 | 5 | 1165 | ||
JN709865.1 | 5 | 1207 | ||
JN709866.1 | 4 | 1123 | ||
JN709867.1 | 5 | 1207 | ||
JN709868.1 | 5 | 1249 | ||
JN709869.1 | 4 | 1165 | ||
L08850.1 | 6 | 1231 | AAA16117.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1012406 | Chr.4:89246630 - 90245636 on Build GRCh38 | Loss | MMRN1 GPRIN3 CCSER1 SNCA-AS1 SNCA |
nsv594806 | Chr.4:88621130 - 90539537 on Build GRCh38 | Gain | FAM13A NAP1L5 MMRN1 TIGD2 GPRIN3 CCSER1 SNCA-AS1 FAM13A-AS1 SNCA HERC3 |
Set Membership: |
Intragenic Exonic DGV Variation |