Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 113235 |
Gene Name: | solute carrier family 46 member 1 |
Gene Aliases: |
G21, HCP1, PCFT |
Location: |
Chr.17:28394642-28406630 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC46A1 | NM_001242366.2 | NP_001229295.1 | ||
NM_080669.5 | NP_542400.2 | |||
XM_005277786.3 | XP_005277843.1 | |||
XM_017024110.1 | XP_016879599.1 | |||
AK054669.1 | BAB70789.1 | |||
AK074161.1 | BAB84987.1 | |||
AK097194.1 | 1 | 301 | ||
AK295883.1 | 2 | 1116 | ||
AL832613.2 | CAD89945.1 | |||
BC010691.1 | AAH10691.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1062056 | Chr.17:28376191 - 28452023 on Build GRCh38 | Gain | SLC46A1 SARM1 |
nsv1056839 | Chr.17:28348199 - 28428188 on Build GRCh38 | Gain | SLC46A1 SARM1 SEBOX POLDIP2 MIR4723 TMEM199 VTN |
nsv574653 | Chr.17:28227749 - 28501479 on Build GRCh38 | Gain | PYY2 IFT20 SEBOX MIR4723 LOC105371712 LOC105371930 SLC46A1 RPS7P1 KRT18P55 SARM1 TNFAIP1 TMEM97 POLDIP2 PPY2P TMEM199 LOC101926984 SLC13A2 VTN |
Set Membership: |
Intragenic Non-exonic DGV Variation |