Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 28952 |
Gene Name: | coiled-coil domain containing 22 |
Gene Aliases: |
CXorf37, JM1, RTSC2 |
Location: |
Chr.X:49235467-49250526 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC22 | NM_014008.4 | NP_054727.1 | ||
XM_005272599.3 | XP_005272656.1 | |||
AJ005890.1 | CAA06747.1 | |||
AK291976.1 | ||||
AK298264.1 | ||||
AK316375.1 | ||||
BC000972.2 | AAH00972.1 | |||
BC011675.2 | AAH11675.1 |
Entrez Gene ID: | 50943 |
Gene Name: | forkhead box P3 |
Gene Aliases: |
AIID, DIETER, IPEX, JM2, PIDX, XPID |
Location: |
Chr.X:49250436-49266505 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOXP3 | NM_001114377.1 | 11 | 2166 | NP_001107849.1 |
NM_014009.3 | 12 | 2271 | NP_054728.2 | |
XM_006724533.2 | 11 | 2446 | XP_006724596.2 | |
XM_011543916.2 | 11 | 3876 | XP_011542218.1 | |
XM_017029565.1 | 13 | 3732 | XP_016885054.1 | |
XM_017029566.1 | 12 | 3696 | XP_016885055.1 | |
XM_017029567.1 | 10 | 2271 | XP_016885056.1 | |
BQ184335.1 | 1 | 127 | ||
DB342786.1 | 1 | 109 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | Chr.X:49205142 - 49320261 on Build GRCh38 | Gain | GAGE10 CACNA1F CCDC22 PPP1R3F FOXP3 |
nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain | SLC38A5 CCDC120 PCSK1N FTSJ1 SLC35A2 MAGIX FOXP3 RBM3 PQBP1 GLOD5 KCND1 SYP-AS1 SYP WDR13 GPKOW LOC101927635 PRICKLE3 PORCN OTUD5 PLP2 PIM2 SUV39H1 CACNA1F CCDC22 TFE3 TBC1D25 WDR45 WAS PRAF2 TIMM17B HDAC6 PPP1R3F GRIPAP1 ERAS EBP GATA1 |
esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss | SSX1 SLC38A5 CCDC120 PCSK1N FTSJ1 SSX4B SLC35A2 MAGIX SSX9 FOXP3 RBM3 PQBP1 GLOD5 SSX3 KCND1 ZNF630 SYP-AS1 SYP SSX6 WDR13 GPKOW LOC101927635 ZNF182 PRICKLE3 PORCN SSX4 OTUD5 ZNF630-AS1 PLP2 PIM2 SPACA5 SUV39H1 CACNA1F CCDC22 TFE3 TBC1D25 WDR45 WAS PRAF2 TIMM17B SPACA5B HDAC6 SSX5 PPP1R3F GRIPAP1 ERAS EBP GATA1 |
nsv469734 | Chr.X:49205141 - 49320261 on Build GRCh38 | Gain+Loss | GAGE10 CACNA1F CCDC22 PPP1R3F FOXP3 |
nsv6895 | Chr.X:49227319 - 49262690 on Build GRCh38 | Insertion | CACNA1F CCDC22 FOXP3 |
Set Membership: |
Intragenic Exonic Non-exonic DGV Variation |