Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2346 |
Gene Name: | folate hydrolase (prostate-specific membrane antigen) 1 |
Gene Aliases: |
FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP |
Location: |
Chr.11:49145094-49208706 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv467874 | Chr.11:48483044 - 49935211 on Build GRCh38 | Gain | FOLH1 TRIM64C TRIM53CP LOC440040 OR4A47 TRIM49B TRIM51CP |
nsv554347 | Chr.11:49089915 - 49935211 on Build GRCh38 | Gain | FOLH1 LOC440040 |
nsv518571 | Chr.11:49141396 - 49606281 on Build GRCh38 | Gain | FOLH1 LOC440040 |
esv2759825 | Chr.11:49190685 - 49447414 on Build GRCh38 | Gain+Loss | FOLH1 |
esv3891996 | Chr.11:49173538 - 49443281 on Build GRCh38 | Gain | FOLH1 |
nsv1044070 | Chr.11:49126191 - 49427222 on Build GRCh38 | Loss | FOLH1 |
esv3891995 | Chr.11:49134487 - 49221694 on Build GRCh38 | Gain | FOLH1 |
esv2744485 | Chr.11:48902336 - 49894003 on Build GRCh38 | Deletion | FOLH1 TRIM64C TRIM53CP LOC440040 TRIM49B TRIM51CP |
nsv972015 | Chr.11:49165970 - 49219167 on Build GRCh38 | Duplication | FOLH1 |
nsv469956 | Chr.11:48901093 - 49846333 on Build GRCh38 | Gain | FOLH1 TRIM64C TRIM53CP LOC440040 TRIM49B TRIM51CP |
esv3891986 | Chr.11:48021501 - 49282806 on Build GRCh38 | Gain | FOLH1 TRIM64C OR4X2 TRIM53CP OR4A47 OR4C3 OR4B1 OR4X1 TRIM49B PTPRJ MIR3161 OR4S1 TRIM51CP |
esv3891984 | Chr.11:48711661 - 49369067 on Build GRCh38 | Gain | FOLH1 TRIM64C TRIM53CP TRIM49B TRIM51CP |
esv2666171 | Chr.11:48963976 - 49887121 on Build GRCh38 | Deletion | FOLH1 TRIM64C TRIM53CP LOC440040 TRIM49B |
Set Membership: |
Intragenic Exonic DGV Variation |