Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3172 |
Gene Name: | hepatocyte nuclear factor 4 alpha |
Gene Aliases: |
FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14 |
Location: |
Chr.20:44355801-44432845 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HNF4A | NM_000457.4 | 10 | 2607 | NP_000448.3 |
NM_001030003.2 | 10 | 2398 | NP_001025174.1 | |
NM_001258355.1 | 11 | 2698 | NP_001245284.1 | |
NM_001287182.1 | 11 | 2620 | NP_001274111.1 | |
NM_001287183.1 | 11 | 2650 | NP_001274112.1 | |
NM_175914.4 | 10 | 2428 | NP_787110.2 | |
NM_178849.2 | 10 | 2577 | NP_849180.1 | |
XM_005260407.3 | 11 | 2727 | XP_005260464.1 | |
AW935533.1 | 1 | 206 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss | FITM2 HNF4A-AS1 LOC101927200 PTPRT MIR3646 JPH2 LOC101927138 HNF4A OSER1-AS1 SRSF6 OSER1 SGK2 GDAP1L1 IFT52 MYBL2 LOC105372626 R3HDML L3MBTL1 LOC101927159 TOX2 GTSF1L |
nsv1061719 | Chr.20:44415722 - 44602319 on Build GRCh38 | Gain | LINC01430 TTPAL LINC01620 SERINC3 HNF4A PKIG |
nsv586065 | Chr.20:44261867 - 44505244 on Build GRCh38 | Gain | FITM2 LINC01430 TTPAL LINC01620 GDAP1L1 HNF4A-AS1 MIR3646 R3HDML SERINC3 HNF4A |
Set Membership: |
Intragenic Exonic DGV Variation |