Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3559 |
Gene Name: | interleukin 2 receptor subunit alpha |
Gene Aliases: |
CD25, IDDM10, IL2R, IMD41, TCGFR, p55 |
Location: |
Chr.10:6010694-6062370 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL2RA | NM_000417.2 | 8 | 1761 | NP_000408.1 |
NM_001308242.1 | 7 | 1545 | NP_001295171.1 | |
NM_001308243.1 | 6 | 1473 | NP_001295172.1 | |
K03122.1 | 7 | 1485 | AAB59535.1 | |
X01057.1 | 8 | 1722 | CAA25525.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1e49 | Chr.10:5684858 - 6183224 on Build GRCh38 | Loss | RBM17 FAM208B MIR3155A PFKFB3 MIR3155B GDI2 ANKRD16 FBXO18 IL15RA IL2RA |
Set Membership: |
Intragenic Exonic DGV Variation |