Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 84464 |
Gene Name: | SLX4 structure-specific endonuclease subunit |
Gene Aliases: |
BTBD12, FANCP, MUS312 |
Location: |
Chr.16:3581181-3611598 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLX4 | NM_032444.2 | 4 | 1483 | NP_115820.2 |
XM_011522715.2 | 4 | 1497 | XP_011521017.1 | |
AB075867.1 | 3 | 1151 | BAB85573.1 | |
AK122775.1 | 4 | 1484 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833128 | Chr.16:3525122 - 3690497 on Build GRCh38 | Loss | DNASE1 CLUAP1 TRAP1 LOC101929732 SLX4 NLRC3 |
nsv518509 | Chr.16:3583656 - 3688960 on Build GRCh38 | Gain | DNASE1 TRAP1 SLX4 |
nsv517451 | Chr.16:3549655 - 3611814 on Build GRCh38 | Loss | DNASE1 LOC101929732 SLX4 NLRC3 |
Set Membership: |
Intragenic Exonic DGV Variation |