Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2189 |
Gene Name: | Fanconi anemia complementation group G |
Gene Aliases: |
FAG, XRCC9 |
Location: |
Chr.9:35073838-35080016 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FANCG | NM_004629.1 | 7 | 1334 | NP_004620.1 |
AJ007669.1 | 7 | 1334 | CAA07602.1 | |
AK293427.1 | 8 | 1024 | ||
AK311348.1 | 6 | 808 | ||
AK312987.1 | 7 | 1259 | ||
BC000032.2 | 7 | 980 | AAH00032.1 | |
BC011623.2 | 7 | 1107 | AAH11623.1 | |
BT009813.1 | 7 | 842 | AAP88815.1 | |
KU159422.1 | 7 | 1365 | ||
KU178053.1 | 7 | 842 | ||
KU178054.1 | 7 | 842 | ||
KU178055.1 | 7 | 842 | ||
KU178056.1 | 4 | 416 | ||
KU178057.1 | ||||
U70310.1 | 7 | 976 | AAB80802.1 |
Set Membership: |
Intragenic Exonic non-DGV Variation |