Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 54897 |
Gene Name: | castor zinc finger 1 |
Gene Aliases: |
CAS11, CST, SRG, ZNF693, dJ734G22.1 |
Location: |
Chr.1:10636602-10796676 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CASZ1 | NM_001079843.2 | NP_001073312.1 | ||
NM_017766.4 | NP_060236.3 | |||
XM_017001540.1 | XP_016857029.1 | |||
XM_017001541.1 | XP_016857030.1 | |||
AK092289.1 | ||||
DQ217660.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4n27 | Chr.1:10613137 - 10736809 on Build GRCh38 | Loss | CASZ1 PEX14 |
dgv105n54 | Chr.1:10613037 - 10736809 on Build GRCh38 | Loss | CASZ1 PEX14 |
nsv545404 | Chr.1:10697142 - 10736809 on Build GRCh38 | Loss | CASZ1 |
esv29966 | Chr.1:10616743 - 10814095 on Build GRCh38 | Loss | CASZ1 PEX14 |
nsv832314 | Chr.1:10644754 - 10799862 on Build GRCh38 | Loss | CASZ1 |
esv2758921 | Chr.1:10682445 - 10995090 on Build GRCh38 | Loss | C1orf127 CASZ1 |
nsv545400 | Chr.1:10624041 - 10781964 on Build GRCh38 | Loss | CASZ1 PEX14 |
nsv525693 | Chr.1:10634068 - 10780848 on Build GRCh38 | Loss | CASZ1 |
nsv828030 | Chr.1:10617505 - 10735104 on Build GRCh38 | Gain | CASZ1 PEX14 |
nsv953454 | Chr.1:10616244 - 10724643 on Build GRCh38 | Deletion | CASZ1 PEX14 |
nsv508858 | Chr.1:10650567 - 10753153 on Build GRCh38 | Insertion | CASZ1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |