Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03863000_cn
          See other LAMP1 CNV Assays ›
          Gene Symbol
          LAMP1
          Assay Reference Genome
          Location

          Chr.13:113313160 on build GRCh38
          Cytoband
          13q34
          Assay ID Hs03863000_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          3916

          Gene Name:

          lysosomal associated membrane protein 1

          Gene Aliases:

          CD107a, LAMPA, LGP120

          Location:

          Chr.13:113297154-113323431 on Build GRCh38

          Assay Gene Location:

          Within Intron 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          LAMP1 NM_005561.3 NP_005552.3
          XM_011537494.1 XP_011535796.1
          AB209319.1 BAD92556.1
          AK092398.1
          AK301584.1
          BC006345.2 AAH06345.2
          BC007845.2 AAH07845.2
          BC021288.2 AAH21288.1
          BC093044.1 AAH93044.1
          BU849686.1
          J03263.1 AAA59524.1
          J04182.1 AAA60382.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv951927 Chr.13:113312286 - 113380185 on Build GRCh38 Deletion LAMP1 LOC105370378 GRTP1-AS1 GRTP1
          esv3633598 Chr.13:113310386 - 113324953 on Build GRCh38 Loss LAMP1 GRTP1
          nsv1048814 Chr.13:113081728 - 113611496 on Build GRCh38 Gain CUL4A MIR8075 ADPRHL1 TMCO3 F10 F7 PROZ F10-AS1 PCID2 MCF2L DCUN1D2 TFDP1 LOC102724474 LAMP1 LOC101928841 LOC105370378 GRTP1-AS1 GRTP1
          nsv509521 Chr.13:113308960 - 113394480 on Build GRCh38 Insertion LAMP1 LOC105370378 GRTP1-AS1 GRTP1
          nsv563316 Chr.13:113261171 - 113347907 on Build GRCh38 Loss CUL4A MIR8075 LAMP1 LOC105370378 GRTP1
          nsv563317 Chr.13:113292168 - 113369086 on Build GRCh38 Loss LAMP1 LOC105370378 GRTP1-AS1 GRTP1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagy
          granzyme-mediated apoptotic signaling pathway
          positive regulation of natural killer cell degranulation
          positive regulation of natural killer cell mediated cytotoxicity
          viral entry into host cell
          autophagic cell death
          protein stabilization
          establishment of protein localization to organelle
          Golgi to lysosome transport
          regulation of organelle transport along microtubule
          virus receptor activity
          protein binding
          enzyme binding
          protein domain specific binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0