Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 5579 |
Gene Name: | protein kinase C beta |
Gene Aliases: |
PKC-beta, PKCB, PRKCB1, PRKCB2 |
Location: |
Chr.16:23835979-24220611 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRKCB | NM_002738.6 | NP_002729.2 | ||
NM_212535.2 | NP_997700.1 | |||
BC036472.1 | AAH36472.1 | |||
M13975.1 | AAA60095.1 | |||
X06318.1 | CAA29634.1 | |||
X07109.1 | CAA30130.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833174 | Chr.16:24077853 - 24258848 on Build GRCh38 | Gain | MIR1273H PRKCB CACNG3 |
esv34056 | Chr.16:24123791 - 24318980 on Build GRCh38 | Loss | MIR1273H PRKCB CACNG3 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |