Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3990 |
Gene Name: | lipase C, hepatic type |
Gene Aliases: |
HDLCQ12, HL, HTGL, LIPH |
Location: |
Chr.15:58410754-58568952 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPC | NM_000236.2 | NP_000227.2 | ||
XM_005254372.1 | XP_005254429.1 | |||
AK290062.1 | ||||
AK292631.1 | ||||
AK293324.1 | ||||
AK309448.1 | ||||
AK315306.1 | ||||
BC132825.1 | ||||
BC136495.1 | ||||
BC146659.1 | ||||
D83548.1 | BAA12014.1 | |||
J03540.1 | AAA59520.1 | |||
J03895.1 | AAA61165.1 | |||
X07228.1 | CAA30188.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv569598 | Chr.15:58260407 - 58560264 on Build GRCh38 | Loss | LOC101928668 LIPC LIPC-AS1 LOC101928635 |
nsv569601 | Chr.15:58508567 - 58573957 on Build GRCh38 | Loss | LOC101928668 LIPC |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |