Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 5579 |
Gene Name: | protein kinase C beta |
Gene Aliases: |
PKC-beta, PKCB, PRKCB1, PRKCB2 |
Location: |
Chr.16:23835979-24220611 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRKCB | NM_002738.6 | NP_002729.2 | ||
NM_212535.2 | NP_997700.1 | |||
BC036472.1 | AAH36472.1 | |||
BI549165.1 | ||||
DB456157.1 | ||||
M13975.1 | AAA60095.1 | |||
X06318.1 | CAA29634.1 | |||
X07109.1 | CAA30130.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3553249 | Chr.16:23922680 - 23931179 on Build GRCh38 | Deletion | PRKCB |
esv23662 | Chr.16:23922584 - 23930911 on Build GRCh38 | Loss | PRKCB |
dgv4958n54 | Chr.16:23922978 - 23928626 on Build GRCh38 | Loss | PRKCB |
esv3638240 | Chr.16:23922814 - 23930927 on Build GRCh38 | Loss | PRKCB |
dgv808e212 | Chr.16:23922513 - 23931242 on Build GRCh38 | Loss | PRKCB |
esv2662823 | Chr.16:23922808 - 23930929 on Build GRCh38 | Deletion | PRKCB |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |