Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 65018 |
Gene Name: | PTEN induced putative kinase 1 |
Gene Aliases: |
BRPK, PARK6 |
Location: |
Chr.1:20633455-20651511 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PINK1 | NM_032409.2 | NP_115785.1 | ||
AB053323.1 | BAB55647.1 | |||
AF316873.1 | AAK28062.1 | |||
AK130746.1 | ||||
BC028215.1 | AAH28215.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain | SH2D5 DDOST KIF17 PINK1-AS PINK1 PLA2G2F CAMK2N1 MIR6084 PLA2G2C MUL1 LINC01141 UBXN10-AS1 HP1BP3 PLA2G2D VWA5B1 UBXN10 CDA FAM43B PLA2G5 |
nsv834369 | Chr.1:20609770 - 20779837 on Build GRCh38 | Loss | SH2D5 MIR6084 DDOST KIF17 HP1BP3 PINK1-AS PINK1 CDA |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |