Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 79628 |
Gene Name: | SH3 domain and tetratricopeptide repeats 2 |
Gene Aliases: |
CMT4C, MNMN |
Location: |
Chr.5:148982150-149063174 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SH3TC2 | NM_024577.3 | NP_078853.2 | ||
AB075865.1 | BAB85571.1 | |||
AK127248.1 | BAC86899.1 | |||
AK300420.1 | ||||
AL832651.2 | ||||
AY341075.1 | AAR03497.1 | |||
BC113879.1 | ||||
BC114486.1 | ||||
DA856608.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830517 | Chr.5:148953659 - 149171578 on Build GRCh38 | Gain | MIR584 SH3TC2 LOC255187 ABLIM3 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |