Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3636 |
Gene Name: | inositol polyphosphate phosphatase like 1 |
Gene Aliases: |
OPSMD, SHIP2 |
Location: |
Chr.11:72223861-72239147 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
INPPL1 | XM_005273978.3 | 1 | 103 | XP_005274035.1 |
XM_005273979.4 | XP_005274036.1 | |||
XM_011544999.2 | XP_011543301.1 | |||
XM_011545000.2 | XP_011543302.1 | |||
XM_017017673.1 | XP_016873162.1 |
Set Membership: |
Intragenic Exonic non-DGV Variation |