Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 128178 |
Gene Name: | EDAR associated death domain |
Gene Aliases: |
ECTD11A, ECTD11B, ED3, EDA3 |
Location: |
Chr.1:236394380-236484708 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EDARADD | NM_080738.3 | NP_542776.1 | ||
NM_145861.2 | NP_665860.2 | |||
AK096339.1 | ||||
AK290862.1 | ||||
AK291930.1 | ||||
AK314634.1 | ||||
AY028913.1 | AAK40287.1 | |||
AY028914.1 | AAK40288.1 | |||
AY071862.1 | AAL60590.1 | |||
BC128082.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1005877 | Chr.1:235912335 - 237038508 on Build GRCh38 | Gain | GPR137B MT1HL1 NID1 ACTN2 ERO1B HEATR1 MTR LGALS8 EDARADD LGALS8-AS1 |
esv2763410 | Chr.1:236334208 - 236735742 on Build GRCh38 | Gain | ACTN2 HEATR1 LGALS8 EDARADD LGALS8-AS1 |
dgv591n100 | Chr.1:236442533 - 236626827 on Build GRCh38 | Gain | HEATR1 LGALS8 EDARADD LGALS8-AS1 |
esv2762227 | Chr.1:236440911 - 236623173 on Build GRCh38 | Gain | HEATR1 LGALS8 EDARADD LGALS8-AS1 |
nsv516855 | Chr.1:236435808 - 236624489 on Build GRCh38 | Gain | HEATR1 LGALS8 EDARADD LGALS8-AS1 |
nsv549394 | Chr.1:235876577 - 236629706 on Build GRCh38 | Gain | GPR137B NID1 ERO1B HEATR1 LYST LGALS8 EDARADD LGALS8-AS1 |
nsv1012436 | Chr.1:235860657 - 236648100 on Build GRCh38 | Loss | GPR137B NID1 ERO1B HEATR1 LYST LGALS8 EDARADD LGALS8-AS1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |