Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 1298 |
Gene Name: | collagen type IX alpha 2 |
Gene Aliases: |
DJ39G22.4, EDM2, MED, STL5 |
Location: |
Chr.1:40300487-40317653 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COL9A2 | NM_001852.3 | NP_001843.1 | ||
XM_006710365.3 | XP_006710428.1 | |||
XM_011540715.2 | XP_011539017.1 | |||
XM_011540716.2 | XP_011539018.1 | |||
XM_011540717.2 | XP_011539019.1 | |||
XM_017000332.1 | XP_016855821.1 | |||
XM_017000333.1 | XP_016855822.1 | |||
BC136326.1 | ||||
BC136327.1 | ||||
M95610.1 | AAA80977.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv950687 | Chr.1:40294329 - 40321928 on Build GRCh38 | Deletion | COL9A2 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |