Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2904 |
Gene Name: | glutamate ionotropic receptor NMDA type subunit 2B |
Gene Aliases: |
EIEE27, GluN2B, MRD6, NMDAR2B, NR2B, hNR3 |
Location: |
Chr.12:13537337-13982012 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRIN2B | NM_000834.3 | NP_000825.2 | ||
XM_011520628.2 | XP_011518930.1 | |||
XM_011520629.2 | XP_011518931.1 | |||
XM_017019219.1 | XP_016874708.1 | |||
AB208850.1 | BAD92087.1 | |||
BC113618.1 | ||||
BC113620.1 | ||||
U11287.1 | AAB60368.1 | |||
U88963.1 | AAD00659.1 | |||
U90278.1 | AAB49993.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv826262 | Chr.12:13632554 - 13725802 on Build GRCh38 | Loss | GRIN2B |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |