Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4625 |
Gene Name: | myosin, heavy chain 7, cardiac muscle, beta |
Gene Aliases: |
CMD1S, CMH1, MPD1, MYHCB, SPMD, SPMM |
Location: |
Chr.14:23412738-23435686 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYH7 | NM_000257.3 | NP_000248.2 | ||
XM_017021340.1 | XP_016876829.1 | |||
AB209708.1 | BAD92945.1 | |||
AK307590.1 | ||||
AY518538.1 | AAT44534.1 | |||
BC112171.1 | AAI12172.1 | |||
BC112173.1 | AAI12174.1 | |||
EF560725.1 | ||||
EU747717.1 | ||||
M58018.1 | AAA62830.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv974315 | Chr.14:23425338 - 23426796 on Build GRCh38 | Duplication | MYH7 |
esv3584149 | Chr.14:23395407 - 23427942 on Build GRCh38 | Gain | MYH7 MHRT MIR208B MYH6 |
Set Membership: |
Intragenic Non-exonic DGV Variation |