Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 57716 |
Gene Name: | periaxin |
Gene Aliases: |
CMT4F |
Location: |
Chr.19:40393762-40414718 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRX | NM_020956.2 | NP_066007.1 | ||
NM_181882.2 | NP_870998.2 | |||
XM_011527171.2 | XP_011525473.1 | |||
AF321191.1 | AAK19279.1 | |||
AF321192.1 | AAK19280.1 | |||
BC067266.1 | AAH67266.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833831 | Chr.19:40290833 - 40454958 on Build GRCh38 | Loss | PRX MIR6796 C19orf47 SERTAD3 HIPK4 SERTAD1 PLD3 BLVRB LOC105372489 |
nsv953575 | Chr.19:40404294 - 40416093 on Build GRCh38 | Deletion | PRX |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |