Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3172 |
Gene Name: | hepatocyte nuclear factor 4 alpha |
Gene Aliases: |
FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14 |
Location: |
Chr.20:44355801-44432845 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HNF4A | NM_000457.4 | NP_000448.3 | ||
NM_001030003.2 | NP_001025174.1 | |||
NM_001030004.2 | NP_001025175.1 | |||
NM_001258355.1 | NP_001245284.1 | |||
NM_001287182.1 | NP_001274111.1 | |||
NM_001287183.1 | NP_001274112.1 | |||
NM_001287184.1 | NP_001274113.1 | |||
NM_175914.4 | NP_787110.2 | |||
NM_178849.2 | NP_849180.1 | |||
NM_178850.2 | NP_849181.1 | |||
XM_005260407.3 | XP_005260464.1 | |||
AB307703.1 | ||||
AY680696.1 | AAT91237.1 | |||
AY680697.1 | AAT91238.1 | |||
AY680698.1 | AAT91239.1 | |||
BC137539.1 | ||||
BC137540.1 | ||||
BP274413.1 | ||||
FJ608822.1 | ||||
HQ692860.1 | ||||
HQ692868.1 | ||||
HQ692869.1 | ||||
X76930.1 | CAA54248.1 | |||
X87870.1 | CAA61133.1 | |||
X87871.1 | CAA61134.1 | |||
X87872.1 | CAA61135.1 | |||
Z49825.1 | CAA89989.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss | OSER1 PTPRT SRSF6 TOX2 LOC101927138 HNF4A MYBL2 R3HDML LOC105372626 LOC101927159 IFT52 LOC101927200 GDAP1L1 JPH2 OSER1-AS1 L3MBTL1 FITM2 MIR3646 SGK2 HNF4A-AS1 GTSF1L |
nsv586065 | Chr.20:44261867 - 44505244 on Build GRCh38 | Gain | LINC01620 GDAP1L1 FITM2 SERINC3 HNF4A TTPAL R3HDML LINC01430 MIR3646 HNF4A-AS1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |