Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 15211 |
Gene Name: | hexosaminidase A |
Gene Aliases: |
Hex-1 |
Location: |
Chr.9:59539545-59565109 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hexa | NM_010421.4 | NP_034551.2 | ||
AK075895.1 | BAC36036.1 | |||
AK075911.1 | BAC36049.1 | |||
AK080777.1 | BAC38018.1 | |||
AK144168.1 | BAE25744.1 | |||
AK150444.1 | BAE29566.1 | |||
AK151960.1 | BAE30831.1 | |||
AK159091.1 | BAE34808.1 | |||
AK159814.1 | BAE35394.1 | |||
AK159890.1 | BAE35457.1 | |||
AK160112.1 | BAE35636.1 | |||
AK167013.1 | BAE39189.1 | |||
AK168186.1 | BAE40146.1 | |||
AK170903.1 | ||||
BC010755.1 | AAH10755.1 | |||
U07631.1 | AAA18775.1 | |||
X64331.1 | CAA45615.1 |
Set Membership: |
Intragenic Non-exonic |