Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 66725 |
Gene Name: | leucine-rich repeat kinase 2 |
Gene Aliases: |
4921513O20Rik, 9330188B09Rik, AW561911, D630001M17Rik, Gm927, cI-46 |
Location: |
Chr.15:91672821-91816124 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrrk2 | NM_025730.3 | 28 | 3921 | NP_080006.3 |
AK034413.1 | 5 | 545 | BAC28700.1 | |
AY792512.1 | 28 | 3864 | AAV63976.1 |
Set Membership: |
Intragenic Exonic |