Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CCR7
Transcript:
Assay Name:
Assay ID
CRISPR1125061_LV
Size
200 µL,1x10⁸ TU/mL
200 µL,1x10⁸ TU/mL
Availability
Made To Order
Catalog #
A32042
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
Gene Ontology Categories:
Function(s)
Process(es)
establishment of T cell polarity
positive regulation of cell-matrix adhesion
dendritic cell chemotaxis
myeloid dendritic cell chemotaxis
positive regulation of dendritic cell antigen processing and presentation
positive regulation of hypersensitivity
positive regulation of humoral immune response
inflammatory response
immune response
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
positive regulation of actin filament polymerization
positive regulation of pseudopodium assembly
ruffle organization
response to lipopolysaccharide
regulation of interferon-gamma production
positive regulation of interleukin-12 production
response to prostaglandin E
chemokine (C-C motif) ligand 19 signaling pathway
chemokine (C-C motif) ligand 21 signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of phosphatidylinositol 3-kinase activity
negative thymic T cell selection
positive regulation of cell adhesion
positive regulation of protein kinase activity
positive regulation of JNK cascade
homeostasis of number of cells
regulation of interleukin-1 beta secretion
positive regulation of T cell receptor signaling pathway
release of sequestered calcium ion into cytosol
positive regulation of filopodium assembly
positive regulation of protein kinase B signaling
positive regulation of ERK1 and ERK2 cascade
cellular response to cytokine stimulus
response to nitric oxide
interleukin-12 secretion
positive regulation of neutrophil chemotaxis
activation of GTPase activity
lymphocyte migration into lymph node
mature conventional dendritic cell differentiation
negative regulation of leukocyte apoptotic process
positive regulation of cell motility
positive regulation of dendritic cell chemotaxis
positive regulation of immunological synapse formation
positive regulation of T cell costimulation
positive regulation of glycoprotein biosynthetic process involved in immunological synapse formation
regulation of dendritic cell dendrite assembly
G-protein coupled receptor activity
C-C chemokine receptor activity
chemokine (C-C motif) ligand 19 binding
chemokine (C-C motif) ligand 21 binding
C-C motif chemokine 19 receptor activity
C-C motif chemokine 21 receptor activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-6dgdk:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0