Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PTGER4
Transcript:
Assay Name:
Assay ID
CRISPR1125593_LV
Size
200 µL,1x10⁸ TU/mL
200 µL,1x10⁸ TU/mL
Availability
Made To Order
Catalog #
A32042
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
receptor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of peptide secretion
immune response
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
JNK cascade
female pregnancy
positive regulation of cell proliferation
response to mechanical stimulus
positive regulation of gene expression
negative regulation of hydrogen peroxide metabolic process
regulation of circadian sleep/wake cycle, wakefulness
positive regulation of smooth muscle cell migration
positive regulation of cAMP biosynthetic process
response to lipopolysaccharide
response to progesterone
positive regulation of interleukin-8 production
negative regulation of integrin activation
positive regulation of urine volume
positive regulation of renal sodium excretion
T-helper cell differentiation
negative regulation of circadian sleep/wake cycle, REM sleep
chemokinesis
response to drug
positive regulation of tyrosine phosphorylation of Stat3 protein
response to amino acid
positive regulation of osteoblast differentiation
positive regulation of bone resorption
positive regulation of cell adhesion
positive regulation of circadian sleep/wake cycle, non-REM sleep
negative regulation of cytokine secretion
negative regulation of interleukin-1 alpha secretion
positive regulation of cytokine secretion
negative regulation of inflammatory response
positive regulation of inflammatory response
regulation of stress fiber assembly
negative regulation of nitric-oxide synthase biosynthetic process
maternal process involved in parturition
bone development
positive regulation of mucus secretion
ERK1 and ERK2 cascade
cellular response to mechanical stimulus
cellular response to glucose stimulus
cellular response to interleukin-1
cellular response to prostaglandin E stimulus
positive regulation of wound healing
ductus arteriosus closure
positive regulation of hyaluronan biosynthetic process
response to salt
negative regulation of ductus arteriosus closure
negative regulation of small intestine smooth muscle contraction
positive regulation of calcitonin secretion
positive regulation of chemokinesis
positive regulation of matrix metallopeptidase secretion
negative regulation of tumor necrosis factor secretion
negative regulation of endothelin secretion
positive regulation of substance P secretion, neurotransmission
response to water-immersion restraint stress
positive regulation of antral ovarian follicle growth
positive regulation of neutrophil extravasation
negative regulation of eosinophil extravasation
positive regulation of interleukin-10 secretion
prostaglandin E receptor activity
protein binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-8dfkb:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0