Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Igf1
Transcript:
Assay Name:
Assay ID
CRISPR452589_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell activation
regulation of protein phosphorylation
blood vessel remodeling
regulation of translation
nervous system development
memory
cell proliferation
positive regulation of cell proliferation
negative regulation of cell proliferation
response to heat
glial cell differentiation
regulation of gene expression
positive regulation of cardiac muscle hypertrophy
phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
muscle hypertrophy
myotube cell development
positive regulation of smooth muscle cell migration
positive regulation of cerebellar granule cell precursor proliferation
water homeostasis
proteoglycan biosynthetic process
positive regulation of cell growth
lung development
positive regulation of cell migration
mammary gland development
exocrine pancreas development
regulation of establishment or maintenance of cell polarity
regulation of intracellular steroid hormone receptor signaling pathway
positive regulation of protein import into nucleus, translocation
negative regulation of smooth muscle cell apoptotic process
multicellular organism growth
bone mineralization involved in bone maturation
positive regulation of activated T cell proliferation
regulation of cell proliferation
positive regulation of tyrosine phosphorylation of Stat5 protein
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of DNA binding
positive regulation of MAPK cascade
protein kinase B signaling
positive regulation of insulin-like growth factor receptor signaling pathway
myoblast differentiation
positive regulation of fat cell differentiation
positive regulation of osteoblast differentiation
positive regulation of glycogen biosynthetic process
positive regulation of DNA replication
positive regulation of glycolytic process
positive regulation of mitotic nuclear division
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of glucose import
positive regulation of Ras protein signal transduction
insulin-like growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
positive regulation of fibroblast proliferation
lung alveolus development
cell development
positive regulation of smooth muscle cell proliferation
branching morphogenesis of an epithelial tube
inner ear development
chondroitin sulfate proteoglycan biosynthetic process
positive regulation of epithelial cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
detection of mechanical stimulus involved in sensory perception
regulation of protein metabolic process
myoblast proliferation
positive regulation of protein kinase B signaling
regulation of calcium ion transport
negative regulation of oocyte development
lung vasculature development
lung lobe morphogenesis
Type I pneumocyte differentiation
Type II pneumocyte differentiation
prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
prostate gland growth
prostate gland epithelium morphogenesis
prostate gland stromal morphogenesis
negative regulation of androgen receptor signaling pathway
ERK1 and ERK2 cascade
negative regulation of ERK1 and ERK2 cascade
positive regulation of calcineurin-NFAT signaling cascade
positive regulation of protein serine/threonine kinase activity
positive regulation of steroid hormone biosynthetic process
negative regulation of release of cytochrome c from mitochondria
extrinsic apoptotic signaling pathway in absence of ligand
negative regulation of neuron death
positive regulation of trophectodermal cell proliferation
positive regulation of myoblast proliferation
negative regulation of extrinsic apoptotic signaling pathway
insulin receptor binding
insulin-like growth factor receptor binding
integrin binding
hormone activity
steroid binding
protein binding
growth factor activity
protein serine/threonine kinase activator activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-2px8h:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0