Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Ntrk1
Transcript:
Assay Name:
Assay ID
CRISPR483858_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
multicellular organism development
nervous system development
axon guidance
learning or memory
programmed cell death involved in cell development
response to activity
phosphorylation
peptidyl-tyrosine phosphorylation
sensory perception of pain
cell differentiation
B cell differentiation
response to nutrient levels
response to nicotine
nerve growth factor signaling pathway
mechanoreceptor differentiation
response to drug
positive regulation of programmed cell death
negative regulation of neuron apoptotic process
positive regulation of angiogenesis
protein autophosphorylation
neurotrophin TRK receptor signaling pathway
sympathetic nervous system development
detection of temperature stimulus involved in sensory perception of pain
detection of mechanical stimulus involved in sensory perception of pain
positive regulation of NF-kappaB transcription factor activity
response to electrical stimulus
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
Sertoli cell development
innervation
axonogenesis involved in innervation
behavioral response to formalin induced pain
cellular response to nicotine
cellular response to growth factor stimulus
negative regulation of neuron death
cellular response to nerve growth factor stimulus
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
neurotrophin receptor activity
neurotrophin p75 receptor binding
protein binding
ATP binding
nerve growth factor receptor activity
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
kinase binding
protein homodimerization activity
ephrin receptor binding
nerve growth factor binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-bsnw9:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0