Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Ntf3
Transcript:
Assay Name:
Assay ID
CRISPR514993_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
neurotrophic factor
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
positive regulation of receptor internalization
transmembrane receptor protein tyrosine kinase signaling pathway
neuromuscular synaptic transmission
nervous system development
glial cell fate determination
axon guidance
brain development
peripheral nervous system development
positive regulation of cell proliferation
epidermis development
nerve development
positive regulation of cell migration
activation of protein kinase B activity
positive regulation of peptidyl-serine phosphorylation
mechanoreceptor differentiation
myelination
regulation of apoptotic process
regulation of neuron apoptotic process
negative regulation of neuron apoptotic process
regulation of neuron differentiation
positive regulation of glial cell differentiation
positive regulation of transcription from RNA polymerase II promoter
enteric nervous system development
neuron development
generation of neurons
neuron projection morphogenesis
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of peptidyl-tyrosine phosphorylation
modulation of synaptic transmission
positive chemotaxis
induction of positive chemotaxis
smooth muscle cell differentiation
activation of GTPase activity
positive regulation of actin cytoskeleton reorganization
receptor binding
neurotrophin receptor binding
neurotrophin p75 receptor binding
protein binding
growth factor activity
chemoattractant activity
nerve growth factor binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-2px8h:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0