Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
MAPK8
Transcript:
Assay Name:
Assay ID
CRISPR557642_LV
Size
200 µL,1x10⁸ TU/mL
200 µL,1x10⁸ TU/mL
Availability
Made To Order
Catalog #
A32042
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
response to stress
JNK cascade
JUN phosphorylation
response to UV
positive regulation of gene expression
regulation of macroautophagy
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
regulation of histone deacetylation
positive regulation of cyclase activity
negative regulation of protein binding
regulation of protein localization
Fc-epsilon receptor signaling pathway
regulation of circadian rhythm
positive regulation of apoptotic process
negative regulation of apoptotic process
rhythmic process
regulation of sequence-specific DNA binding transcription factor activity
positive regulation of protein metabolic process
cellular response to lipopolysaccharide
cellular response to mechanical stimulus
positive regulation of deacetylase activity
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
protein serine/threonine kinase activity
JUN kinase activity
protein binding
ATP binding
kinase activity
enzyme binding
histone deacetylase regulator activity
histone deacetylase binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-p89b7:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0