Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
APP
Transcript:
Assay Name:
Assay ID
CRISPR614116_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
response to yeast
suckling behavior
platelet degranulation
mRNA polyadenylation
regulation of translation
protein phosphorylation
cellular copper ion homeostasis
endocytosis
response to oxidative stress
cell adhesion
regulation of epidermal growth factor-activated receptor activity
Notch signaling pathway
axonogenesis
mating behavior
locomotory behavior
axo-dendritic transport
cholesterol metabolic process
adult locomotory behavior
visual learning
cellular process
response to lead ion
negative regulation of endopeptidase activity
positive regulation of peptidase activity
positive regulation of G2/M transition of mitotic cell cycle
axon midline choice point recognition
neuron remodeling
dendrite development
antibacterial humoral response
antifungal humoral response
extracellular matrix organization
forebrain development
neuron projection development
ionotropic glutamate receptor signaling pathway
regulation of multicellular organism growth
regulation of protein binding
cellular protein metabolic process
innate immune response
negative regulation of neuron differentiation
positive regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
collateral sprouting in absence of injury
regulation of synapse structure or activity
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
neuromuscular process controlling balance
synaptic growth at neuromuscular junction
neuron apoptotic process
smooth endoplasmic reticulum calcium ion homeostasis
cellular response to cAMP
cellular response to norepinephrine stimulus
amyloid fibril formation
cellular response to nerve growth factor stimulus
DNA binding
serine-type endopeptidase inhibitor activity
receptor binding
protein binding
heparin binding
peptidase activator activity
enzyme binding
identical protein binding
transition metal ion binding
PTB domain binding
growth factor receptor binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-tvqvm:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0