Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ACE
Transcript:
Assay Name:
Assay ID
CRISPR641411_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
kidney development
blood vessel remodeling
angiotensin maturation
angiotensin catabolic process in blood
regulation of renal output by angiotensin
neutrophil mediated immunity
antigen processing and presentation of peptide antigen via MHC class I
regulation of systemic arterial blood pressure by renin-angiotensin
spermatogenesis
regulation of blood pressure
regulation of smooth muscle cell migration
regulation of vasoconstriction
mononuclear cell proliferation
regulation of vasodilation
hormone catabolic process
peptide catabolic process
beta-amyloid metabolic process
arachidonic acid secretion
heart contraction
regulation of angiotensin metabolic process
hematopoietic stem cell differentiation
positive regulation of protein tyrosine kinase activity
cell proliferation in bone marrow
positive regulation of peptidyl-tyrosine autophosphorylation
regulation of hematopoietic stem cell proliferation
negative regulation of gap junction assembly
positive regulation of peptidyl-cysteine S-nitrosylation
endopeptidase activity
carboxypeptidase activity
drug binding
metallopeptidase activity
exopeptidase activity
tripeptidyl-peptidase activity
peptidyl-dipeptidase activity
zinc ion binding
chloride ion binding
mitogen-activated protein kinase kinase binding
bradykinin receptor binding
mitogen-activated protein kinase binding
metallodipeptidase activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0