Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
HTRA2
Transcript:
Assay Name:
Assay ID
CRISPR796246_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
hydrolase
protease
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
ceramide metabolic process
mitochondrion organization
adult walking behavior
intrinsic apoptotic signaling pathway in response to DNA damage
response to herbicide
positive regulation of cell death
protein autoprocessing
pentacyclic triterpenoid metabolic process
forebrain development
cellular response to oxidative stress
cellular response to heat
cellular response to interferon-beta
regulation of multicellular organism growth
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
cellular protein catabolic process
negative regulation of cell cycle
neuron development
cellular response to retinoic acid
cellular response to growth factor stimulus
execution phase of apoptosis
negative regulation of neuron death
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
regulation of mitophagy
positive regulation of protein targeting to mitochondrion
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
serine-type endopeptidase activity
protein binding
peptidase activity
serine-type peptidase activity
unfolded protein binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-gzxth:80/10.239.166.29:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0