Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ACVRL1
Transcript:
Assay Name:
Assay ID
CRISPR890517_LV
Size
200 µL,1x10⁸ TU/mL
200 µL,1x10⁸ TU/mL
Availability
Made To Order
Catalog #
A32042
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
in utero embryonic development
regulation of endothelial cell proliferation
negative regulation of endothelial cell proliferation
positive regulation of endothelial cell proliferation
lymphangiogenesis
blood vessel maturation
blood vessel remodeling
blood vessel endothelial cell proliferation involved in sprouting angiogenesis
endocardial cushion morphogenesis
regulation of DNA replication
regulation of transcription, DNA-templated
protein phosphorylation
negative regulation of cell adhesion
signal transduction
transforming growth factor beta receptor signaling pathway
blood circulation
regulation of blood pressure
negative regulation of cell proliferation
negative regulation of endothelial cell migration
negative regulation of gene expression
positive regulation of pathway-restricted SMAD protein phosphorylation
signal transduction by protein phosphorylation
negative regulation of cell growth
negative regulation of cell migration
BMP signaling pathway
positive regulation of BMP signaling pathway
positive regulation of chondrocyte differentiation
activin receptor signaling pathway
wound healing, spreading of epidermal cells
dorsal aorta morphogenesis
regulation of blood vessel endothelial cell migration
negative regulation of blood vessel endothelial cell migration
negative regulation of endothelial cell differentiation
positive regulation of endothelial cell differentiation
positive regulation of angiogenesis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein heterooligomerization
negative regulation of focal adhesion assembly
lymphatic endothelial cell differentiation
artery development
venous blood vessel development
endothelial tube morphogenesis
retina vasculature development in camera-type eye
cellular response to transforming growth factor beta stimulus
cellular response to BMP stimulus
endocardial cushion to mesenchymal transition
negative regulation of DNA biosynthetic process
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
transforming growth factor beta-activated receptor activity
transforming growth factor beta receptor activity, type I
protein binding
ATP binding
activin receptor activity, type I
protein kinase binding
SMAD binding
metal ion binding
activin binding
transforming growth factor beta binding
BMP receptor activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0