Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
GSTP1
Transcript:
Assay Name:
Assay ID
CRISPR919386_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
negative regulation of acute inflammatory response
negative regulation of protein kinase activity
glutathione metabolic process
xenobiotic metabolic process
central nervous system development
negative regulation of biosynthetic process
negative regulation of tumor necrosis factor-mediated signaling pathway
oligodendrocyte development
organ regeneration
response to estradiol
negative regulation of interleukin-1 beta production
negative regulation of tumor necrosis factor production
cellular response to insulin stimulus
regulation of stress-activated MAPK cascade
negative regulation of stress-activated MAPK cascade
positive regulation of superoxide anion generation
response to L-ascorbic acid
common myeloid progenitor cell proliferation
nitric oxide storage
negative regulation of apoptotic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
response to amino acid
negative regulation of MAP kinase activity
negative regulation of MAPK cascade
negative regulation of JUN kinase activity
linoleic acid metabolic process
response to ethanol
response to mercury ion
negative regulation of fibroblast proliferation
negative regulation of nitric-oxide synthase biosynthetic process
regulation of ERK1 and ERK2 cascade
negative regulation of ERK1 and ERK2 cascade
negative regulation of leukocyte proliferation
cellular response to lipopolysaccharide
cellular response to epidermal growth factor stimulus
cellular response to glucocorticoid stimulus
cellular response to cell-matrix adhesion
negative regulation of monocyte chemotactic protein-1 production
cellular oxidant detoxification
glutathione derivative biosynthetic process
negative regulation of extrinsic apoptotic signaling pathway
glutathione transferase activity
glutathione peroxidase activity
protein binding
drug binding
JUN kinase binding
kinase regulator activity
S-nitrosoglutathione binding
dinitrosyl-iron complex binding
glutathione binding
nitric oxide binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-5svh6:80/10.239.165.170:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0