Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
FGFR1
Transcript:
Assay Name:
Assay ID
CRISPR923759_LV
Size
200 µL,1x10⁸ TU/mL
200 µL,1x10⁸ TU/mL
Availability
Made To Order
Catalog #
A32042
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
skeletal system development
angiogenesis
ureteric bud development
in utero embryonic development
organ induction
neuron migration
positive regulation of mesenchymal cell proliferation
chondrocyte differentiation
transcription, DNA-templated
protein phosphorylation
sensory perception of sound
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
positive regulation of phospholipase activity
positive regulation of phospholipase C activity
positive regulation of neuron projection development
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
cell migration
peptidyl-tyrosine phosphorylation
stem cell population maintenance
motogenic signaling involved in postnatal olfactory bulb interneuron migration
ventricular zone neuroblast division
embryonic limb morphogenesis
midbrain development
neuron projection development
fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
phosphatidylinositol-3-phosphate biosynthetic process
inner ear morphogenesis
outer ear morphogenesis
middle ear morphogenesis
chordate embryonic development
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of GTPase activity
regulation of cell differentiation
positive regulation of neuron differentiation
negative regulation of osteoblast differentiation
positive regulation of cell cycle
protein autophosphorylation
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
paraxial mesoderm development
regulation of lateral mesodermal cell fate specification
cell maturation
skeletal system morphogenesis
mesenchymal cell differentiation
regulation of sensory perception of pain
positive regulation of cardiac muscle cell proliferation
auditory receptor cell development
branching involved in salivary gland morphogenesis
lung-associated mesenchyme development
regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
regulation of stem cell proliferation
positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
positive regulation of endothelial cell chemotaxis to fibroblast growth factor
regulation of extrinsic apoptotic signaling pathway in absence of ligand
glycoprotein binding
protein tyrosine kinase activity
fibroblast growth factor-activated receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
ATP binding
heparin binding
1-phosphatidylinositol-3-kinase activity
fibroblast growth factor binding
protein complex binding
identical protein binding
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
cell adhesion molecule binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-zbtnl:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0