Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
FGF10
Transcript:
Assay Name:
Assay ID
CRISPR935323_LV
Size
200 µL,1x10⁸ TU/mL
200 µL,1x10⁸ TU/mL
Availability
Made To Order
Catalog #
A32042
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
establishment of mitotic spindle orientation
MAPK cascade
activation of MAPK activity
angiogenesis
metanephros development
organ induction
mesonephros development
blood vessel remodeling
metanephros morphogenesis
determination of left/right symmetry
salivary gland development
negative regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
regulation of smoothened signaling pathway
embryonic pattern specification
epithelial cell migration
positive regulation of epithelial cell migration
positive regulation of keratinocyte proliferation
regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
pituitary gland development
embryonic genitalia morphogenesis
thyroid gland development
otic vesicle formation
positive regulation of vascular endothelial growth factor receptor signaling pathway
pancreas development
hair follicle morphogenesis
embryonic camera-type eye development
actin cytoskeleton reorganization
positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
response to estradiol
response to lipopolysaccharide
positive regulation of ATPase activity
lacrimal gland development
regulation of activin receptor signaling pathway
protein localization to cell surface
somatic stem cell population maintenance
phosphatidylinositol-3-phosphate biosynthetic process
wound healing
tissue regeneration
odontogenesis of dentin-containing tooth
muscle cell fate commitment
positive regulation of MAPK cascade
positive regulation of GTPase activity
keratinocyte proliferation
negative regulation of cell differentiation
positive regulation of DNA repair
positive regulation of DNA replication
positive regulation of Notch signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
positive regulation of Ras protein signal transduction
phosphatidylinositol phosphorylation
regulation of saliva secretion
phosphatidylinositol-mediated signaling
positive regulation of fibroblast proliferation
spleen development
thymus development
embryonic digestive tract morphogenesis
branching morphogenesis of an epithelial tube
female genitalia morphogenesis
male genitalia morphogenesis
positive regulation of lymphocyte proliferation
epithelial cell proliferation
urothelial cell proliferation
positive regulation of urothelial cell proliferation
positive regulation of epithelial cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
white fat cell differentiation
positive chemotaxis
induction of positive chemotaxis
smooth muscle cell differentiation
positive regulation of keratinocyte migration
radial glial cell differentiation
positive regulation of epithelial cell proliferation involved in wound healing
limb bud formation
lung epithelium development
lung saccule development
bronchiole morphogenesis
bud outgrowth involved in lung branching
bud elongation involved in lung branching
mesenchymal-epithelial cell signaling involved in lung development
Type II pneumocyte differentiation
prostatic bud formation
fibroblast growth factor receptor signaling pathway involved in mammary gland specification
mammary gland bud formation
submandibular salivary gland formation
epithelial cell proliferation involved in salivary gland morphogenesis
regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
branch elongation involved in salivary gland morphogenesis
semicircular canal fusion
mesenchymal cell differentiation involved in lung development
secretion by lung epithelial cell involved in lung growth
lung proximal/distal axis specification
tear secretion
positive regulation of white fat cell proliferation
ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
Harderian gland development
negative regulation of cell cycle arrest
positive regulation of hair follicle cell proliferation
positive regulation of canonical Wnt signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
fibroblast growth factor receptor binding
type 2 fibroblast growth factor receptor binding
protein binding
growth factor activity
heparin binding
1-phosphatidylinositol-3-kinase activity
chemoattractant activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-zctgw:80/10.239.163.131:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0