Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ZAP70
Transcript:
Assay Name:
Assay ID
CRISPR941110_LV
Size
200 µL,1x10⁸ TU/mL
200 µL,1x10⁸ TU/mL
Availability
Made To Order
Catalog #
A32042
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
macrophage activation involved in immune response
neutrophil activation involved in immune response
protein phosphorylation
inflammatory response
immune response
transmembrane receptor protein tyrosine kinase signaling pathway
peptidyl-tyrosine phosphorylation
T cell differentiation
positive regulation of cell adhesion mediated by integrin
intracellular signal transduction
peptidyl-tyrosine autophosphorylation
T cell activation
B cell activation
positive regulation of mast cell degranulation
beta selection
positive thymic T cell selection
negative thymic T cell selection
innate immune response
positive regulation of B cell differentiation
positive regulation of T cell differentiation
positive regulation of alpha-beta T cell differentiation
positive regulation of alpha-beta T cell proliferation
positive regulation of calcium-mediated signaling
T cell receptor signaling pathway
B cell receptor signaling pathway
T cell aggregation
T cell migration
regulation of platelet aggregation
phosphotyrosine binding
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-6dgdk:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0