Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CCL5
Transcript:
Assay Name:
Assay ID
CRISPR982604_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
chemokine
cytokine
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
dendritic cell chemotaxis
chronic inflammatory response
monocyte chemotaxis
regulation of chronic inflammatory response
calcium ion transport
cellular calcium ion homeostasis
exocytosis
chemotaxis
inflammatory response
leukocyte cell-cell adhesion
G-protein coupled receptor signaling pathway
cell-cell signaling
aging
response to virus
response to toxic substance
response to salt stress
positive regulation of activation of JAK2 kinase activity
positive regulation of macrophage chemotaxis
positive regulation of T cell chemotaxis
positive regulation of phosphatidylinositol 3-kinase signaling
response to activity
positive regulation of smooth muscle cell migration
dibenzo-p-dioxin metabolic process
positive regulation of cell migration
neutrophil chemotaxis
positive regulation of cellular biosynthetic process
activation of phospholipase D activity
positive regulation of fever generation
lipopolysaccharide-mediated signaling pathway
response to insulin
positive regulation of cell-cell adhesion mediated by integrin
positive regulation of homotypic cell-cell adhesion
positive regulation of T cell proliferation
neutrophil activation
positive regulation of phosphorylation
response to drug
positive regulation of tyrosine phosphorylation of STAT protein
protein kinase B signaling
positive regulation of GTPase activity
cellular protein complex assembly
response to estrogen
negative regulation by host of viral transcription
cellular response to fibroblast growth factor stimulus
positive regulation of viral genome replication
negative regulation of viral genome replication
positive regulation of innate immune response
positive regulation of neuron differentiation
positive regulation of osteoclast differentiation
negative regulation of G-protein coupled receptor protein signaling pathway
positive regulation of angiogenesis
positive regulation of cell adhesion
positive regulation of translational initiation
positive regulation of JAK-STAT cascade
eosinophil chemotaxis
macrophage chemotaxis
lymphocyte chemotaxis
positive regulation of smooth muscle cell proliferation
positive regulation of epithelial cell proliferation
positive regulation of inflammatory response
regulation of insulin secretion
regulation of T cell activation
positive chemotaxis
protein tetramerization
response to glucocorticoid
positive regulation of calcium ion transport
positive regulation of mast cell chemotaxis
positive regulation of protein tyrosine kinase activity
chemokine-mediated signaling pathway
negative regulation of chemokine-mediated signaling pathway
negative regulation of T cell apoptotic process
positive regulation of T cell apoptotic process
positive regulation of ERK1 and ERK2 cascade
response to cholesterol
cellular response to amino acid stimulus
cellular response to vitamin K
cellular response to morphine
cellular response to interferon-gamma
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to ethanol
cellular response to high density lipoprotein particle stimulus
cellular response to organic cyclic compound
cellular response to alkyl hydroperoxide
cellular response to transforming growth factor beta stimulus
positive regulation of monocyte chemotaxis
regulation of neuron death
negative regulation of neuron death
negative regulation of macrophage apoptotic process
positive regulation of T cell migration
positive regulation of natural killer cell chemotaxis
phosphatidylinositol phospholipase C activity
protein kinase activity
protein binding
chemokine activity
heparin binding
phospholipase activator activity
receptor signaling protein tyrosine kinase activator activity
CCR1 chemokine receptor binding
CCR4 chemokine receptor binding
CCR5 chemokine receptor binding
chemoattractant activity
chemokine receptor binding
protein homodimerization activity
protein self-association
chemokine receptor antagonist activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kqhf4:80/10.239.162.79:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0