Search
Search

| Assay Name: | gga-miR-6581-3p |
|---|---|
| miRBase Accession Number: | MI0022400 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUCUCCCUGGUGGGCUUUUU |
| Species: | Chicken |
| Product Type: | QuantiGene Singleplex miRNA |
Mature miRNA Sequence: | UGUCUCCCUGGUGGGCUUUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chicken | gga-miR-6581-3p | MIMAT0025670 |