Search
Search

CHGA| Gene Symbol: CHGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001275.4 | NP_001266.1 | ||||
| NM_001301690.2 | NP_001288619.1 | |||||
| XM_011536370.3 | XP_011534672.1 | |||||
| GenBank mRNA | AK223381.1 | - | ||||
| AK313757.1 | - | |||||
| BC001059.2 | - | |||||
| BC006459.1 | - | |||||
| BC009384.2 | - | |||||
| BC012755.2 | - | |||||
| BT006869.1 | - | |||||
| BX248263.1 | - | |||||
| DA201149.1 | - | |||||
| J03483.1 | - | |||||
| J03915.1 | - | |||||