Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_024105.3 | NP_077010.1 | ||||
XM_011530371.2 | XP_011528673.1 | |||||
XM_017028936.1 | XP_016884425.1 | |||||
XM_017028937.1 | XP_016884426.1 | |||||
GenBank mRNA | AF258571.1 | - | ||||
AF318343.1 | - | |||||
AJ290427.1 | - | |||||
AJ303120.1 | - | |||||
BC001729.1 | - | 3353085 | ||||
BC098562.1 | - | 30560020 | ||||
BM145575.1 | - | |||||
CB858085.1 | - | |||||
CR456369.1 | - | |||||
ENST00000330817.0 | - |