Search
Search

ORAI2| Gene Symbol: ORAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001126340.2 | NP_001119812.1 | ||||
| NM_001271818.1 | NP_001258747.1 | |||||
| NM_001271819.1 | NP_001258748.1 | |||||
| NM_032831.3 | NP_116220.1 | |||||
| GenBank mRNA | AF258552.1 | - | ||||
| AK022536.1 | - | |||||
| AK027639.1 | - | |||||
| AK056497.1 | - | |||||
| AK128753.1 | - | |||||
| AK300574.1 | - | |||||
| BC009183.2 | - | |||||
| BC016797.1 | - | |||||
| BC069270.1 | - | 5477442 | ||||
| CN366874.1 | - | |||||
| CR457287.1 | - | |||||
| DA770788.1 | - | |||||
| DC384892.1 | - | |||||
| ENST00000356387.0 | - | |||||
| HY064081.1 | - | |||||
| Y10777.1 | - | |||||