Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001010.2 | NP_001001.2 | ||||
GenBank mRNA | AB062123.1 | - | ||||
AK093634.1 | - | |||||
AK291517.1 | - | |||||
AK311861.1 | - | |||||
BC000524.2 | - | 2961137 | ||||
BC009427.2 | - | |||||
BC013296.2 | - | 3030319 | ||||
BC027620.1 | - | 4698843 | ||||
BC035447.1 | - | |||||
BC071907.1 | - | 4900361 | ||||
BC071908.1 | - | 5089456 | ||||
BC094826.1 | - | 5481378 | ||||
D28348.1 | - | |||||
ENST00000380394.0 | - | |||||
J03537.1 | - | |||||
M20020.1 | - |