Search
Search

IFIH1| Gene Symbol: IFIH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_022168.4 | NP_071451.2 | ||||
| XM_047445407.1 | XP_047301363.1 | |||||
| GenBank mRNA | AF095844.1 | - | ||||
| AK056293.1 | - | |||||
| AK292941.1 | - | |||||
| AK314611.1 | - | |||||
| AY017378.1 | - | |||||
| BC007966.2 | - | |||||
| BC046208.1 | - | |||||
| BC078180.1 | - | |||||
| BC111750.1 | - | |||||
| BQ772836.1 | - | |||||
| BU902097.1 | - | |||||