Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TARP EBGEX Assays ›
Gene Symbol:
TARP
Gene Name:
TCR gamma alternate reading frame protein
Gene Aliases:
CD3G
Chromosome Location:
Chr.7: 38259643 - 38273636 on Build GRCh38
UniGene ID:
Hs.534032
Species:
Human
Species Specific ID (Flybase ID):
-
Type:
1
Detection Label:
Alexa Fluor 647
ProductType:
PrimeFlow Probe Set
Assay ID
VA1-19674-PF
Size
40 tests
100 tests
Availability
Made To Order
Catalog #
PF-204
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: TARP
Interrogated Sequence
Translated Protein
IMAGE Clone ID
RefSeq
NM_001003799.2
NP_001003799.1
NM_001003806.2
NP_001003806.1
GenBank mRNA
AF151103.1
-
AF512447.1
-
AF512448.1
-
AF512449.1
-
AF512450.1
-
AF512451.1
-
AF512452.1
-
AF512453.1
-
AF512454.1
-
AF512455.1
-
AF512456.1
-
AF512457.1
-
AK096766.1
-
BC017861.1
-
BC030554.1
-
BC105589.1
-
BF674593.1
-
EU395807.1
-
JQ956403.1
-
KC170727.1
-
M16804.1
-
M17324.1
-
M17325.1
-
M27331.1
-
M27332.1
-
M27334.1
-
M27335.1
-
M27339.1
-
M27340.1
-
M27341.1
-
M27342.1
-
M27343.1
-
M27344.1
-
M27345.1
-
M30894.1
-
M33028.1
-
M89818.1
-
M89819.1
-
M89820.1
-
M89821.1
-
M89824.1
-
M89827.1
-
M89831.1
-
M89832.1
-
M89833.1
-
M89835.1
-
M89836.1
-
M89837.1
-
M89839.1
-
M89840.1
-
M89842.1
-
M89843.1
-
M89844.1
-
M89845.1
-
M89846.1
-
M89847.1
-
M89848.1
-
M89849.1
-
M89850.1
-
M89851.1
-
M89853.1
-
M89855.1
-
M89856.1
-
S60175.1
-
S60779.1
-
S60780.1
-
S73842.1
-
X06774.1
-
X06775.1
-
X06776.1
-
X72500.1
-
X74774.1
-
X74775.1
-
Y00790.1
-
Z11169.1
-
Z11170.1
-
Z11171.1
-
Z11172.1
-
Z11173.1
-
Z11174.1
-
Z11175.1
-
Z11176.1
-
Z11177.1
-
Z11178.1
-
Z11179.1
-
Z11180.1
-
Z11181.1
-
Z11182.1
-
Z11183.1
-
Z11184.1
-
Z11185.1
-
Z11186.1
-
Z11187.1
-
Z11188.1
-
Z11189.1
-
Z11190.1
-
Z11191.1
-
Z11192.1
-
Z11193.1
-
Z11194.1
-
Z11195.1
-
Z11196.1
-
Z11197.1
-
Z11198.1
-
Z11199.1
-
Z11200.1
-
Z11201.1
-
Z11202.1
-
Z11203.1
-
Z11204.1
-
Z11205.1
-
Z11206.1
-
Z11208.1
-
Z11209.1
-
Z11210.1
-
Z11211.1
-
Z11212.1
-
Z11213.1
-
Z11214.1
-
Z11215.1
-
Z11216.1
-
Z11217.1
-
Z11218.1
-
Z11219.1
-
Z11220.1
-
Z11221.1
-
Z11222.1
-
Z11223.1
-
Z11224.1
-
Z11225.1
-
Z11226.1
-
Z11227.1
-
Z11228.1
-
Z11229.1
-
Z11230.1
-
Z11231.1
-
Z11232.1
-
Z11233.1
-
Z11234.1
-
Z11235.1
-
Z11236.1
-
Z11238.1
-
Z11239.1
-
Z11241.1
-
Z11242.1
-
Z11243.1
-
Z11244.1
-
Z11245.1
-
Z11246.1
-
Z11247.1
-
Z11248.1
-
Z11249.1
-
Z11250.1
-
Z11251.1
-
Z11252.1
-
Z11253.1
-
Z11254.1
-
Z11255.1
-
Z11256.1
-
Z11257.1
-
Z11258.1
-
Z11259.1
-
Z11261.1
-
Z11262.1
-
Z11263.1
-
Z11264.1
-
Z11265.1
-
Z11266.1
-
Z11267.1
-
Z11268.1
-
Z11269.1
-
Z11270.1
-
Z11271.1
-
Z11272.1
-
Z11273.1
-
Z11274.1
-
Z11275.1
-
Z11276.1
-
Z11277.1
-
Z11278.1
-
Z11279.1
-
Z11280.1
-
Z11281.1
-
Z11282.1
-
Z11283.1
-
Z11284.1
-
Z11285.1
-
Z11286.1
-
Z11287.1
-
Z11288.1
-
Z11289.1
-
Z11290.1
-
Z11291.1
-
Z11292.1
-
Z11294.1
-
Z11295.1
-
Z11296.1
-
Z11297.1
-
Z11298.1
-
Z11299.1
-
Z11300.1
-
Z11301.1
-
Z11302.1
-
Z11303.1
-
Z11304.1
-
Z11305.1
-
Z11306.1
-
Z11307.1
-
Z11308.1
-
Z11309.1
-
Z11310.1
-
Z11311.1
-
Z11313.1
-
Z11314.1
-
Z11315.1
-
Z11316.1
-
Z11318.1
-
Z11319.1
-
Z11320.1
-
Z11321.1
-
Z11322.1
-
Z11323.1
-
Z11324.1
-
Z11325.1
-
Z11326.1
-
Z11327.1
-
Z11328.1
-
Z11329.1
-
Z11330.1
-
Z11331.1
-
Z11332.1
-
Z11333.1
-
Z11334.1
-
Z11335.1
-
Z11336.1
-
Z11337.1
-
Z11338.1
-
Z11339.1
-
Z11340.1
-
Z11341.1
-
Z11342.1
-
Z11343.1
-
Z11344.1
-
Z11345.1
-
Z11346.1
-
Z11347.1
-
Z11348.1
-
Z11349.1
-
Z11350.1
-
Z11351.1
-
Z11352.1
-
Z11353.1
-
Z11354.1
-
Z11355.1
-
Z11356.1
-
Z11357.1
-
Z11358.1
-
Z11359.1
-
Z11360.1
-
Z11361.1
-
Z11362.1
-
Z11363.1
-
Z11364.1
-
Z11365.1
-
Z11366.1
-
Z11367.1
-
Z11368.1
-
Z11369.1
-
Z11370.1
-
Z11371.1
-
Z11372.1
-
Z11373.1
-
Z11374.1
-
Z11375.1
-
Z11376.1
-
Z11377.1
-
Z11378.1
-
Z11379.1
-
Z11380.1
-
Z11381.1
-
Z11382.1
-
Z11383.1
-
Z11384.1
-
Z11385.1
-
Z11386.1
-
Z11387.1
-
Z11388.1
-
Z11389.1
-
Z11390.1
-
Z11391.1
-
Z11392.1
-
Z11393.1
-
Z11394.1
-
Z11395.1
-
Z11396.1
-
Z11397.1
-
Z11398.1
-
Z11399.1
-
Z11400.1
-
Z11401.1
-
Z11402.1
-
Z11403.1
-
Z11404.1
-
Z11405.1
-
Z11406.1
-
Z11407.1
-
Z11408.1
-
Z11409.1
-
Z11410.1
-
Back To Top
More Information
Back To Top
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline