Search
Search

CEACAM1| Gene Symbol: CEACAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001024912.3 | NP_001020083.1 | ||||
| NM_001184813.2 | NP_001171742.1 | |||||
| NM_001184815.2 | NP_001171744.1 | |||||
| NM_001184816.2 | NP_001171745.1 | |||||
| NM_001205344.2 | NP_001192273.1 | |||||
| NM_001712.5 | NP_001703.2 | |||||
| XM_011527206.3 | XP_011525508.1 | |||||
| GenBank mRNA | AK314211.1 | - | ||||
| AL833584.1 | - | |||||
| AY766113.1 | - | |||||
| BC014473.1 | - | |||||
| BC024164.1 | - | |||||
| BU161659.1 | - | |||||
| BX485906.1 | - | |||||
| D12502.1 | - | |||||
| D90311.1 | - | |||||
| D90312.1 | - | |||||
| D90313.1 | - | |||||
| DA432598.1 | - | |||||
| J03858.1 | - | |||||
| M69176.1 | - | |||||
| M72238.1 | - | |||||
| M76742.1 | - | |||||
| M76743.1 | - | |||||
| M76744.1 | - | |||||
| S71326.1 | - | |||||
| X14831.1 | - | |||||
| X16354.1 | - | |||||
| X16356.1 | - | |||||