Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001114123.2 | NP_001107595.1 | ||||
NM_001257168.1 | NP_001244097.1 | |||||
NM_005229.4 | NP_005220.2 | |||||
XM_017029339.1 | XP_016884828.1 | |||||
GenBank mRNA | AB016193.1 | - | ||||
AB209047.1 | - | |||||
AF000672.1 | - | |||||
AK093966.1 | - | |||||
AK312984.1 | - | |||||
BC048296.1 | - | |||||
BC056150.1 | - | 6503270 | ||||
BQ448492.1 | - | |||||
CB852603.1 | - | |||||
CR542252.1 | - | |||||
ENST00000247161.0 | - | |||||
M25269.1 | - |