Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: FGF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_002009.3 | NP_002000.1 | ||||
GenBank mRNA | AK313160.1 | - | ||||
BC010956.1 | - | 4282925 | ||||
CR542002.1 | - | |||||
DB206350.1 | - | |||||
ENST00000267843.0 | - | |||||
M60828.1 | - | |||||
S81661.1 | - |