Search
Search

ITPR1| Gene Symbol: ITPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001099952.4 | NP_001093422.2 | ||||
| NM_001168272.2 | NP_001161744.1 | |||||
| NM_001378452.1 | NP_001365381.1 | |||||
| NM_002222.7 | NP_002213.5 | |||||
| GenBank mRNA | AB208868.1 | - | ||||
| AI470158.1 | - | |||||
| AK293752.1 | - | |||||
| AK294594.1 | - | |||||
| AK296235.1 | - | |||||
| AK309981.1 | - | |||||
| AK311008.1 | - | |||||
| AW501800.1 | - | |||||
| BC144563.1 | - | |||||
| BG202086.1 | - | |||||
| BM695661.1 | - | |||||
| BQ006717.1 | - | |||||
| CN431631.1 | - | |||||
| CV355426.1 | - | |||||
| D26070.1 | - | |||||
| DA060305.1 | - | |||||
| DB260851.1 | - | |||||
| DB544988.1 | - | |||||
| DB576768.1 | - | |||||
| L38019.2 | - | |||||
| S82269.1 | - | |||||
| U23850.2 | - | |||||